Search
Search

Sptbn2| Gene Symbol: Sptbn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021287.1 | NP_067262.1 | 30 - 31 | 6674 | 76 | |
| GenBank mRNA | AK147671.1 | - | 30 - 31 | 6690 | 76 | |
| AK172931.1 | - | 20 - 21 | 4983 | 76 | ||
| BC048851.1 | - | 8 - 9 | 1492 | 76 | ||
| BC079628.1 | - | 8 - 9 | 1626 | 76 | ||
| BC079860.1 | - | 30 - 31 | 6674 | 6834745 | 76 | |
Set Membership: |
|