Search
Search

Adrbk1| Gene Symbol: Adrbk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290818.1 | NP_001277747.1 | 6 - 7 | 714 | 129 | |
| NM_130863.2 | NP_570933.1 | 7 - 8 | 786 | 129 | ||
| GenBank mRNA | AF333028.1 | - | 6 - 7 | 506 | 129 | |
| AK155691.1 | - | 6 - 7 | 715 | 129 | ||
| BC003196.1 | - | 6 - 7 | 502 | 129 | ||
| BC053922.1 | - | 7 - 8 | 519 | 6309555 | 129 | |
Set Membership: |
|