Search
Search
| Gene Symbol: Ist1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028018.2 | NP_082294.1 | 3 - 4 | 398 | 64 | |
| GenBank mRNA | AK010262.1 | - | 3 - 4 | 368 | 64 | |
| AK050468.1 | - | 3 - 4 | 365 | 64 | ||
| AK088192.1 | - | 3 - 4 | 353 | 64 | ||
| AK122215.1 | - | 1 - 2 | 172 | 64 | ||
| AK152830.1 | - | 3 - 4 | 365 | 64 | ||
| AK161899.1 | - | 3 - 4 | 354 | 64 | ||
| AK207302.1 | - | 1 - 2 | 92 | 64 | ||
| BC017605.1 | - | 3 - 4 | 364 | 4507347 | 64 | |
| BC039052.1 | - | 3 - 4 | 341 | 2631157 | 64 | |
Set Membership: |
|