Search
Search

Cdc25a| Gene Symbol: Cdc25a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007658.3 | NP_031684.3 | 6 - 7 | 937 | 130 | |
| GenBank mRNA | AK077609.1 | - | 6 - 7 | 554 | 130 | |
| AK077991.1 | - | 6 - 7 | 940 | 130 | ||
| AK142334.1 | - | 5 - 6 | 1212 | 130 | ||
| BC046296.1 | - | 6 - 7 | 556 | 6486343 | 130 | |
Set Membership: |
|