Search
Search

Adamts18| Gene Symbol: Adamts18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172466.2 | NP_766054.2 | 2 - 3 | 548 | 62 | |
| GenBank mRNA | AK035797.1 | - | 2 - 3 | 448 | 62 | |
| AK053851.1 | - | 1 - 2 | 656 | 62 | ||
| AK081122.1 | - | 2 - 3 | 451 | 62 | ||
| AK140876.1 | - | 2 - 3 | 324 | 62 | ||
| BC094674.1 | - | 2 - 3 | 203 | 30548647 | 62 | |
| CB246179.1 | - | 1 - 2 | 197 | 62 | ||
Set Membership: |
|