Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01240573_m1
          See other PLTP GE Assays ›
          Gene Symbol:
          Pltp
          Gene Name:
          phospholipid transfer protein
          Gene Aliases:
          Bpife, OD107
          Chromosome Location:
          Chr.2: 164839518 - 164857708 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01240573_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pltp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011125.2 NP_035255.1 3 - 4 333 64
          GenBank mRNA AK148330.1 - 3 - 4 338 64
          AK149730.1 - 3 - 4 264 64
          BC003782.1 - 3 - 4 267 3491360 64
          CT010317.1 - 2 - 3 197 64
          U28960.1 - 2 - 3 191 64
          U37226.1 - 3 - 4 263 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005136.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pltp

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          transport
          lipid transport
          vitamin E biosynthetic process
          positive regulation of cholesterol efflux
          sperm motility
          high-density lipoprotein particle remodeling
          lipid binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200043796.mm.1 NM_011125.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043796.mm.1 NM_011125.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043796.mm.1 TC0200005136.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200043793.mm.1 NM_011125.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043793.mm.1 NM_011125.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043793.mm.1 TC0200005136.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200043795.mm.1 NM_011125.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043795.mm.1 NM_011125.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043795.mm.1 TC0200005136.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200043791.mm.1 NM_011125.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043791.mm.1 NM_011125.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043791.mm.1 TC0200005136.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200043804.mm.1 NM_011125.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043804.mm.1 NM_011125.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043804.mm.1 TC0200005136.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200043800.mm.1 NM_011125.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043800.mm.1 NM_011125.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200043800.mm.1 TC0200005136.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 598999 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline