Search
Search

Slc38a8| Gene Symbol: Slc38a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009950.1 | NP_001009950.1 | 4 - 5 | 693 | 67 | |
| GenBank mRNA | BC089013.1 | - | 4 - 5 | 693 | 30605563 | 67 |
Set Membership: |
|