Search
Search

Taf1c| Gene Symbol: Taf1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021441.2 | NP_067416.2 | 8 - 9 | 950 | 158 | |
| GenBank mRNA | AK030522.1 | - | 8 - 9 | 917 | 158 | |
| AK052250.1 | - | 8 - 9 | 990 | 158 | ||
| AK087534.1 | - | 8 - 9 | 940 | 158 | ||
| AK154846.1 | - | 8 - 9 | 934 | 158 | ||
| AK156718.1 | - | 8 - 9 | 917 | 158 | ||
| BC051944.1 | - | 8 - 9 | 1002 | 158 | ||
| BC058383.1 | - | 8 - 9 | 892 | 6811470 | 158 | |
| Y09974.1 | - | 8 - 9 | 1077 | 158 | ||
Set Membership: |
|