Search
Search

Dnah17| Gene Symbol: Dnah17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167746.1 | NP_001161218.1 | 66 - 67 | 10686 | 91 | |
| GenBank mRNA | AK173343.1 | - | 24 - 25 | 4113 | 91 | |
Set Membership: |
|