Search
Search

Abca8b| Gene Symbol: Abca8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013851.2 | NP_038879.2 | 25 - 26 | 3634 | 65 | |
| GenBank mRNA | AF498362.1 | - | 25 - 26 | 3634 | 65 | |
| AK173034.1 | - | 24 - 25 | 3375 | 65 | ||
| BC141401.1 | - | 25 - 26 | 3597 | 9056014 | 65 | |
Set Membership: |
|