Search
Search

Eral1| Gene Symbol: Eral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022313.2 | NP_071708.2 | 2 - 3 | 434 | 78 | |
| GenBank mRNA | AB049389.1 | - | 2 - 3 | 410 | 78 | |
| AF190731.1 | - | 2 - 3 | 419 | 78 | ||
| AK012155.1 | - | 2 - 3 | 424 | 78 | ||
| BC037603.1 | - | 2 - 3 | 419 | 78 | ||
| BC068271.1 | - | 2 - 3 | 429 | 30008388 | 78 | |
Set Membership: |
|