Search
Search

Eral1| Gene Symbol: Eral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022313.2 | NP_071708.2 | 7 - 8 | 978 | 88 | |
| GenBank mRNA | AB049389.1 | - | 7 - 8 | 954 | 88 | |
| AK012155.1 | - | 7 - 8 | 968 | 88 | ||
| AK153947.1 | - | 1 - 2 | 1237 | 88 | ||
| BC019728.1 | - | 1 - 2 | 166 | 88 | ||
| BC037603.1 | - | 7 - 8 | 963 | 88 | ||
| BC068271.1 | - | 7 - 8 | 973 | 30008388 | 88 | |
Set Membership: |
|