Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Il15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001254747.1 | NP_001241676.1 | 5 - 6 | 682 | 108 | |
NM_008357.2 | NP_032383.1 | 5 - 6 | 692 | 108 | ||
GenBank mRNA | AK155616.1 | - | 5 - 6 | 1330 | 108 | |
BC023698.1 | - | 5 - 6 | 677 | 5345382 | 108 | |
U14332.1 | - | 5 - 6 | 651 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |