Search
Search

P2rx5| Gene Symbol: P2rx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033321.3 | NP_201578.2 | 2 - 3 | 354 | 62 | |
| GenBank mRNA | AF333331.1 | - | 2 - 3 | 360 | 62 | |
| AK134662.1 | - | 2 - 3 | 314 | 62 | ||
| BC138189.1 | - | 2 - 3 | 354 | 8861208 | 62 | |
| BC145181.1 | - | 2 - 3 | 354 | 9053732 | 62 | |
| JX570808.1 | - | 2 - 3 | 350 | 62 | ||
Set Membership: |
|