Search
Search
| Gene Symbol: Ltf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008522.3 | NP_032548.2 | 11 - 12 | 1384 | 69 | |
| GenBank mRNA | AK036491.1 | - | 11 - 12 | 1385 | 69 | |
| AK144556.1 | - | 11 - 12 | 1385 | 69 | ||
| AK150799.1 | - | 11 - 12 | 1385 | 69 | ||
| AK151822.1 | - | 11 - 12 | 1385 | 69 | ||
| AK164781.1 | - | 11 - 12 | 1386 | 69 | ||
| BC006904.1 | - | 11 - 12 | 1392 | 3600033 | 69 | |
| BC008530.1 | - | 11 - 12 | 1365 | 69 | ||
| BC009662.1 | - | 7 - 8 | 815 | 69 | ||
| CT010339.1 | - | 11 - 12 | 1350 | 69 | ||
| FJ538998.1 | - | 12 - 13 | 1350 | 69 | ||
| J03298.1 | - | 11 - 12 | 1353 | 69 | ||
Set Membership: |
|