Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01243212_m1
          See other CKS1BRT GE Assays ›
          Gene Symbol:
          Cks1brt
          Gene Name:
          CDC28 protein kinase 1b, retrogene
          Gene Aliases:
          EG624855, Gm6531
          Chromosome Location:
          Chr.8: 85151924 - 85173622 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01243212_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cks1brt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037922.3 NP_001033011.1 1 - 2 99 77
          GenBank mRNA AK144293.1 - 1 - 2 100 77

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK144293.1, NM_001037922.3 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2057142 BC147101.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2057142 NM_001037922.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2057142 AK144293.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2057142 BC147100.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070962 NM_001037922.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070962 AK144293.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 926674 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 275563 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 670277 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 391525 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 721328 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 521304 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 262420 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 464658 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 390212 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 475279 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 20340 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 144614 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161451 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070962 NM_001037922.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2070962 AK144293.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2057142 BC147101.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2057142 NM_001037922.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2057142 AK144293.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2057142 BC147100.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023865 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01784766 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01784765 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161451 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161451 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01784766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07422949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06927132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06861071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06863751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07132500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06927130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07267736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023892 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10573611 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573612 - -
          Affymetrix HT MG-430 PM Array Plate 1438600_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10573610 - -
          Mouse Genome 430 2.0 Array 1438600_at - -
          Mouse Genome 430 2.0 Array 1438600_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 624855_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 624855_at - -
          Affymetrix Mouse Clariom S Array PSR0800007587.mm.1 NM_001037922.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007587.mm.1 NM_001037922.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007587.mm.1 TC0800000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007584.mm.1 NM_001037922.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007584.mm.1 NM_001037922.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007584.mm.1 TC0800000984.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007588.mm.1 NM_001037922.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007588.mm.1 NM_001037922.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007588.mm.1 TC0800000984.mm.1 MTA TC
          Mouse Expression Set 430 1438600_at - -
          Mouse Expression Set 430 1438600_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1438600_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1438600_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 592888 - -
          ABI Mouse Genome Survey Microarray 903874 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2916239 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2916239 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline