Search
Search
| Gene Symbol: Nae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144931.3 | NP_659180.1 | 15 - 16 | 1223 | 154 | |
| GenBank mRNA | AK146202.1 | - | 15 - 16 | 1181 | 154 | |
| BC019163.1 | - | 15 - 16 | 1203 | 3711115 | 154 | |
| BC021510.1 | - | 15 - 16 | 1204 | 5347277 | 154 | |
| BC023680.1 | - | 15 - 16 | 1205 | 5343341 | 154 | |
| BC023897.1 | - | 15 - 16 | 1180 | 5321187 | 154 | |
| BC027124.1 | - | 15 - 16 | 1167 | 5355631 | 154 | |
Set Membership: |
|