Search
Search

Cfdp1| Gene Symbol: Cfdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011801.1 | NP_035931.1 | 5 - 6 | 740 | 77 | |
| GenBank mRNA | AB010828.1 | - | 5 - 6 | 740 | 77 | |
| AK133989.1 | - | 5 - 6 | 758 | 77 | ||
| AK160532.1 | - | 5 - 6 | 705 | 77 | ||
| BC005589.1 | - | 5 - 6 | 721 | 3964401 | 77 | |
| Y08219.1 | - | 5 - 6 | 739 | 77 | ||
Set Membership: |
|