Search
Search
| Gene Symbol: Mcm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK033196.1 | - | 7 - 8 | 978 | 62 | |
| AK051795.1 | - | 7 - 8 | 948 | 62 | ||
| AK133826.1 | - | 7 - 8 | 979 | 62 | ||
| BC090645.1 | - | 7 - 8 | 931 | 62 | ||
| BC094416.1 | - | 7 - 8 | 943 | 5707724 | 62 | |
| D26090.1 | - | 6 - 7 | 998 | 62 | ||
Set Membership: |
|