Search
Search
| Gene Symbol: Nkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163660.1 | NP_001157132.1 | 5 - 6 | 565 | 87 | |
| NM_027280.3 | NP_081556.3 | 7 - 8 | 775 | 87 | ||
| GenBank mRNA | AF343352.1 | - | 7 - 8 | 612 | 87 | |
| AF358134.1 | - | 7 - 8 | 691 | 87 | ||
| AK082367.1 | - | 7 - 8 | 764 | 87 | ||
| AK166564.1 | - | 7 - 8 | 778 | 87 | ||
| BC034838.1 | - | 7 - 8 | 749 | 1429943 | 87 | |
| DV662736.1 | - | 5 - 6 | 545 | 87 | ||
Set Membership: |
|