Search
Search
| Gene Symbol: Aars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146217.4 | NP_666329.2 | 9 - 10 | 1666 | 101 | |
| GenBank mRNA | AK036738.1 | - | 5 - 6 | 2850 | 101 | |
| AK044451.1 | - | 9 - 10 | 1417 | 101 | ||
| AK085725.1 | - | 9 - 10 | 1607 | 101 | ||
| AK150227.1 | - | 9 - 10 | 1594 | 101 | ||
| AK158139.1 | - | 9 - 10 | 1615 | 101 | ||
| AY223875.1 | - | 9 - 10 | 1606 | 101 | ||
| BC013261.1 | - | 6 - 7 | 891 | 101 | ||
| BC033273.1 | - | 1541 | 4196789 | 101 | ||
Set Membership: |
|