Search
Search

Als2cl| Gene Symbol: Als2cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146059.1 | NP_001139531.1 | 12 - 13 | 1372 | 109 | |
| NM_001146060.1 | NP_001139532.1 | 12 - 13 | 1452 | 109 | ||
| NM_146228.4 | NP_666340.3 | 12 - 13 | 1369 | 109 | ||
| GenBank mRNA | AB107016.1 | - | 12 - 13 | 1369 | 109 | |
| AF481142.1 | - | 11 - 12 | 1265 | 109 | ||
| AK030815.1 | - | 5 - 6 | 506 | 109 | ||
| AK053076.1 | - | 12 - 13 | 1342 | 109 | ||
| AK131149.1 | - | 12 - 13 | 1371 | 109 | ||
| AK155451.1 | - | 12 - 13 | 1438 | 109 | ||
| BC117928.1 | - | 11 - 12 | 1322 | 40097635 | 109 | |
| BC117929.1 | - | 11 - 12 | 1319 | 40097636 | 109 | |
Set Membership: |
|