Search
Search

Als2cl| Gene Symbol: Als2cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146059.1 | NP_001139531.1 | 19 - 20 | 2227 | 89 | |
| NM_001146060.1 | NP_001139532.1 | 19 - 20 | 2307 | 89 | ||
| NM_146228.4 | NP_666340.3 | 19 - 20 | 2224 | 89 | ||
| GenBank mRNA | AB107016.1 | - | 19 - 20 | 2224 | 89 | |
| AF481142.1 | - | 18 - 19 | 2120 | 89 | ||
| AK053076.1 | - | 19 - 20 | 2197 | 89 | ||
| AK155451.1 | - | 19 - 20 | 2293 | 89 | ||
| BC117928.1 | - | 18 - 19 | 2177 | 40097635 | 89 | |
| BC117929.1 | - | 18 - 19 | 2174 | 40097636 | 89 | |
Set Membership: |
|