Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01245440_m1
          See other SPINK8 GE Assays ›
          Gene Symbol:
          Spink8
          Gene Name:
          serine peptidase inhibitor, Kazal type 8
          Gene Aliases:
          C630041L24Rik
          Chromosome Location:
          Chr.9: 109812203 - 109826628 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01245440_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spink8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183136.2 NP_898959.2 4 - 5 424 134
          GenBank mRNA CN843460.1 - 4 - 5 370 134
          DQ437329.1 - 4 - 5 406 134

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spink8

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of peptidase activity
          serine-type endopeptidase inhibitor activity
          peptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981291 AK021276.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981291 AW455991.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981291 BC048588.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981291 NM_183136.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981291 DQ437329.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 682149 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 266188 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 28367 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 781970 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 420666 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336875 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 521734 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 26595 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151708 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981291 AK021276.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981291 AW455991.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981291 BC048588.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981291 NM_183136.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981291 DQ437329.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26820487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26820492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26820512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26820508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26820511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26820484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26820515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26820491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26820521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26820514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26820519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26820516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26820522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26820517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26820494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26820483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26820478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26820480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26820490 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02027906 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151708 - -
          Whole Mouse Genome G4122A A_51_P458638 AK021276.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P458638 AW455991.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P458638 BC048588.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P458638 CN843460.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P458638 NM_183136.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140520 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151708 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07470065 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07775307 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07470067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26820601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07838871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07931345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07576551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07702317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07775307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07622532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07838868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07470065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07900075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07931347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07900080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26820472 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10589408 - -
          Affymetrix Mouse Gene 1.0 ST Array 10589411 - -
          Affymetrix Mouse Gene 1.0 ST Array 10589410 - -
          Affymetrix Mouse Gene 1.0 ST Array 10589409 - -
          Affymetrix Mouse Gene 1.0 ST Array 10589412 - -
          Affymetrix HT MG-430 PM Array Plate 1435999_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10589407 - -
          Mouse Genome 430 2.0 Array 1435999_at - -
          Mouse Genome 430 2.0 Array 1435999_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78709_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78709_PM_at - -
          Mouse Expression Set 430 1435999_PM_at - -
          Mouse Expression Set 430 1435999_at - -
          Murine Genome U74v2 Set 166451_PM_f_at - -
          Murine Genome U74v2 Set 166451_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1435999_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435999_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 462810 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4670086 - -
          Illumina mouse-6 v1.0 expression beadchip 4670086 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004670086 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1220865 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220865 - -
          mouse-6 v1.1 (Illumina) scl36443.5.1_93-S - -
          Illumina Sentrix mouse-6.v1 bead array 4670086 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36443.5.1_93-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4670086 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1220865 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1220865 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl36443.5.1_93-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline