Search
Search

Nlgn2| Gene Symbol: Nlgn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198862.2 | NP_942562.2 | 5 - 6 | 1447 | 57 | |
| GenBank mRNA | AK136883.1 | - | 6 - 7 | 2013 | 57 | |
| AK173159.1 | - | 5 - 6 | 1447 | 57 | ||
| BE952199.1 | - | 5 - 6 | 651 | 57 | ||
Set Membership: |
|