Search
Search

Pelp1| Gene Symbol: Pelp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029231.4 | NP_083507.3 | 6 - 7 | 721 | 68 | |
| GenBank mRNA | AK005027.1 | - | 6 - 7 | 719 | 68 | |
| AK128966.1 | - | 6 - 7 | 721 | 68 | ||
| AK217016.1 | - | 3 - 4 | 294 | 68 | ||
| BC090620.1 | - | 6 - 7 | 715 | 30699968 | 68 | |
Set Membership: |
|