Search
Search
| Gene Symbol: Srr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163311.1 | NP_001156783.1 | 3 - 4 | 478 | 75 | |
| NM_013761.4 | NP_038789.1 | 4 - 5 | 704 | 75 | ||
| GenBank mRNA | AF148321.1 | - | 3 - 4 | 359 | 75 | |
| AK031687.1 | - | 4 - 5 | 576 | 75 | ||
| AK043738.1 | - | 4 - 5 | 610 | 75 | ||
| AK043819.1 | - | 3 - 4 | 384 | 75 | ||
| AK080830.1 | - | 4 - 5 | 584 | 75 | ||
| AK157122.1 | - | 3 - 4 | 629 | 75 | ||
| AK170096.1 | - | 4 - 5 | 590 | 75 | ||
| AK170471.1 | - | 4 - 5 | 590 | 75 | ||
| BC011164.1 | - | 4 - 5 | 581 | 4195695 | 75 | |
Set Membership: |
|