Search
Search

Ube2g1| Gene Symbol: Ube2g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025985.4 | NP_080261.2 | 3 - 4 | 571 | 94 | |
| GenBank mRNA | AK012539.1 | - | 3 - 4 | 478 | 94 | |
| AK013902.1 | - | 3 - 4 | 452 | 94 | ||
| AK083029.1 | - | 3 - 4 | 514 | 94 | ||
| AK167472.1 | - | 3 - 4 | 437 | 94 | ||
| BC065083.1 | - | 4 - 5 | 665 | 94 | ||
| BC096474.1 | - | 688 | 4503077 | 94 | ||
| DV657669.1 | - | 3 - 4 | 571 | 94 | ||
Set Membership: |
|