Search
Search

Nol11| Gene Symbol: Nol11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161329.1 | NP_001154801.1 | 16 - 17 | 2007 | 83 | |
| NM_133702.2 | NP_598463.2 | 16 - 17 | 2067 | 83 | ||
| GenBank mRNA | AK038696.1 | - | 16 - 17 | 1898 | 83 | |
| AK076704.1 | - | 16 - 17 | 2008 | 83 | ||
| AK078524.1 | - | 16 - 17 | 1958 | 83 | ||
| AK157743.1 | - | 16 - 17 | 2007 | 83 | ||
| BC004785.1 | - | 11 - 12 | 1346 | 83 | ||
| BC024874.1 | - | 16 - 17 | 1946 | 3984027 | 83 | |
| BC079576.1 | - | 2 - 3 | 1101 | 83 | ||
Set Membership: |
|