Search
Search
| Gene Symbol: Shpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029031.3 | NP_083307.1 | 3 - 4 | 532 | 62 | |
| GenBank mRNA | AK015273.1 | - | 3 - 4 | 555 | 62 | |
| AK171027.1 | - | 4 - 5 | 612 | 62 | ||
| BC054721.1 | - | 3 - 4 | 532 | 5694588 | 62 | |
| BC056433.1 | - | 3 - 4 | 532 | 5694588 | 62 | |
Set Membership: |
|