Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Nedd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008682.2 | NP_032708.2 | 4 - 5 | 449 | 106 | |
GenBank mRNA | AK010591.1 | - | 4 - 5 | 437 | 106 | |
AK048393.1 | - | 4 - 5 | 401 | 106 | ||
AK085355.1 | - | 4 - 5 | 412 | 106 | ||
AK090040.1 | - | 4 - 5 | 394 | 106 | ||
AK153746.1 | - | 4 - 5 | 451 | 106 | ||
BC052430.1 | - | 4 - 5 | 418 | 6836583 | 106 | |
BC066870.1 | - | 4 - 5 | 386 | 30476383 | 106 | |
D10712.1 | - | 4 - 5 | 391 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |