Search
Search

Trhde| Gene Symbol: Trhde | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146241.2 | NP_666353.1 | 2 - 3 | 1128 | 77 | |
| GenBank mRNA | AK029833.1 | - | 457 | 77 | ||
| AK034093.1 | - | 2 - 3 | 507 | 77 | ||
| AK163726.1 | - | 2066 | 77 | |||
| BC032288.1 | - | 2 - 3 | 1133 | 5368580 | 77 | |
Set Membership: |
|