Search
Search

Acadvl| Gene Symbol: Acadvl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017366.3 | NP_059062.1 | 4 - 5 | 376 | 77 | |
| GenBank mRNA | AK076037.1 | - | 4 - 5 | 337 | 77 | |
| AU067294.1 | - | 4 - 5 | 376 | 77 | ||
| BC026559.1 | - | 4 - 5 | 355 | 5037878 | 77 | |
| Z71189.1 | - | 4 - 5 | 297 | 77 | ||
Set Membership: |
|