Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01248916_m1
          See other USP36 GE Assays ›
          Gene Symbol:
          Usp36
          Gene Name:
          ubiquitin specific peptidase 36
          Gene Aliases:
          2700002L06Rik, mKIAA1453
          Chromosome Location:
          Chr.11: 118259651 - 118290244 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01248916_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Usp36
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033528.1 NP_001028700.1 15 - 16 2603 106
          GenBank mRNA AK028572.1 - 15 - 16 2649 106
          AK049205.1 - 15 - 16 2433 106
          AK129363.1 - 15 - 16 2401 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100004156.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Protease 3' Most Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          ubiquitin-dependent protein catabolic process
          protein deubiquitination
          proteasome-mediated ubiquitin-dependent protein catabolic process
          regulation of proteasomal protein catabolic process
          regulation of mitophagy
          positive regulation of protein targeting to mitochondrion
          cysteine-type endopeptidase activity
          thiol-dependent ubiquitin-specific protease activity
          peptidase activity
          cysteine-type peptidase activity
          hydrolase activity
          thiol-dependent ubiquitinyl hydrolase activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133767 AK129363.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133767 AK165635.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133767 AK079342.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133767 AK028572.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133767 NM_001033528.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P02321 AK028915.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P02321 NM_001033528.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P627925 AK129363.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P627925 AK049205.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P627925 AK028915.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P627925 AK028572.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P627925 NM_001033528.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P627925 AK129363.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P627925 AK049205.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P627925 AK028915.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P627925 AK028572.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P627925 NM_001033528.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 215187 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674021 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 606230 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 772599 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 139246 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 119766 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 501412 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 641106 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928458 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 38742 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 737850 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537031 AK002372.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537031 AK003073.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537031 AK004150.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537031 BC058123.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537031 XM_901690.2 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537031 XM_903949.2 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537031 XM_922038.2 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 19637 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 449862 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 899399 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323121 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 489032 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 120592 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 30016 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 96355 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 28207 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165550 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164233 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161562 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P02321 AK028915.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P02321 NM_001033528.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133767 AK129363.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133767 AK165635.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133767 AK079342.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133767 AK028572.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133767 NM_001033528.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P627925 AK129363.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P627925 AK049205.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P627925 AK028915.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P627925 AK028572.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P627925 NM_001033528.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P627925 AK129363.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P627925 AK049205.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P627925 AK028915.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P627925 AK028572.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P627925 NM_001033528.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165550 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161562 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164233 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02442239 - -
          Whole Mouse Genome G4122A A_51_P119016 AK049205.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P119016 AK047554.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P119016 AK028572.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P119016 NM_001033528.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P627925 AK129363.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P627925 AK049205.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P627925 AK028915.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P627925 AK028572.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P627925 NM_001033528.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P627925 AK129363.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P627925 AK049205.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P627925 AK028915.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P627925 AK028572.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P627925 NM_001033528.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161562 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165550 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08998551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09594917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09405502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08998541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09092209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08998553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09276684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09725141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09276665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09697251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09594915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08998535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09092207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09276669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09725131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09092202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09725124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08998533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09055458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09405504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09405505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28170405 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10393531 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393515 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393525 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393511 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393512 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393513 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393518 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393526 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393514 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393519 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393516 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393523 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393522 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393527 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393528 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393529 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393524 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393517 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393510 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393521 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393530 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393520 - -
          Affymetrix HT MG-430 PM Array Plate 1458311_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1456094_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1455237_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10393509 - -
          Mouse Genome 430 2.0 Array 1456094_at - -
          Mouse Genome 430 2.0 Array 1455237_at - -
          Mouse Genome 430 2.0 Array 1456094_PM_at - -
          Mouse Genome 430 2.0 Array 1455237_PM_at - -
          Mouse Genome 430 2.0 Array 1458311_at - -
          Mouse Genome 430 2.0 Array 1458311_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72344_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72344_at - -
          Affymetrix Mouse Clariom S Array PSR1100039487.mm.1 NM_001033528.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039487.mm.1 NM_001033528.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039487.mm.1 TC1100004156.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100039492.mm.1 NM_001033528.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039492.mm.1 NM_001033528.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039492.mm.1 TC1100004156.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100039485.mm.1 NM_001033528.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039485.mm.1 NM_001033528.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039485.mm.1 TC1100004156.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100039490.mm.1 NM_001033528.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039490.mm.1 NM_001033528.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039490.mm.1 TC1100004156.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100039486.mm.1 NM_001033528.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039486.mm.1 NM_001033528.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039486.mm.1 TC1100004156.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100039489.mm.1 NM_001033528.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039489.mm.1 NM_001033528.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039489.mm.1 TC1100004156.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100039493.mm.1 NM_001033528.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039493.mm.1 NM_001033528.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100039493.mm.1 TC1100004156.mm.1 MTA TC
          Mouse Expression Set 430 1455237_PM_at - -
          Mouse Expression Set 430 1456094_PM_at - -
          Mouse Expression Set 430 1455237_at - -
          Mouse Expression Set 430 1456094_at - -
          Mouse Expression Set 430 1458311_at - -
          Mouse Expression Set 430 1458311_PM_at - -
          Mouse Genome 430A 2.0 Array 1456094_at - -
          Mouse Genome 430A 2.0 Array 1455237_PM_at - -
          Mouse Genome 430A 2.0 Array 1455237_at - -
          Mouse Genome 430A 2.0 Array 1456094_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1456094_at - -
          Affymetrix HT Mouse Genome 430A Array 1455237_at - -
          Affymetrix HT Mouse Genome 430A Array 1455237_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1456094_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1458311_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1458311_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 557925 - -
          ABI Mouse Genome Survey Microarray 786723 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 100110408 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2491232 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2491232 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244609 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224255 - -
          Illumina Sentrix mouse-6.v1 bead array 100110408 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100110408 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1244609 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline