Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01248999_m1
          See other FENDRR GE Assays ›
          Gene Symbol:
          Fendrr
          Gene Name:
          Foxf1 adjacent non-coding developmental regulatory RNA
          Gene Aliases:
          1110050K14Rik, LL66, linc-Foxf1a, lincFoxf1
          Chromosome Location:
          Chr.8: 121054882 - 121083110 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01248999_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price 434.00
          Your Price
          Online offer: 407.65
          407.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fendrr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK087264.1 - 5 - 6 2210 71
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB534926.2 - 1 - 2 490 Chr. 8 - 121054882 - 121059974 [-] on Build GRCm38 71

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          26878
          Gm26878 -173.7kb Anti-sense Chr. 8 - 120879399 - 120881217 on NCBI Build GRCm38
          Downstream forkhead box F1 Foxf1 AI450827,
          FREAC1,
          Foxf1a,
          Freac-1,
          HFH-8,
          Hfh8
          24.4kb Anti-sense Chr. 8 - 121084386 - 121088154 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 160082 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916117 AK010599.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916117 AK011717.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916117 AK171121.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916117 AK198184.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916117 BC003876.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916117 BC014770.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916117 BC025053.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916117 BC029327.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916117 NM_001038608.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916117 NM_001042408.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916117 NM_178604.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 108885 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 116909 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 754231 D63383.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 754231 D64135.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 754231 NM_011377.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 754231 U40576.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 754231 U42554.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478197 AF144101.2 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478197 AK002754.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478197 AK005080.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478197 AK087390.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478197 AK088888.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478197 AK139288.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478197 AK184184.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478197 BC011087.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 478197 NM_019879.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 464843 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 807321 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 352520 AF373710.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 352520 AK003572.2 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 352520 AK003699.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 352520 AK013852.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 352520 AK085669.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 352520 BC013535.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 352520 NM_025842.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 31911 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901133 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 892463 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 224974 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 385297 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 488997 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 123632 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11258 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161573 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115189 AK087264.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01846049 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161573 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06665893 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102567 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06851127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06920115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07316257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07125906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07125900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06716324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06851110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06851112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07253354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07104640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06946342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07062061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07081473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06886143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06851120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06946358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07347514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06946357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06826227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07062062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07397209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26224167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06916697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06839220 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430012_PM_at - -
          Mouse Genome 430 2.0 Array 1430012_at - -
          Mouse Genome 430 2.0 Array 1430012_PM_at - -
          Mouse Expression Set 430 1430012_PM_at - -
          Mouse Expression Set 430 1430012_at - -
          Affymetrix HT Mouse Genome 430B Array 1430012_at - -
          Affymetrix HT Mouse Genome 430B Array 1430012_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102320086 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline