Search
Search

Cse1l| Gene Symbol: Cse1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023565.3 | NP_076054.1 | 16 - 17 | 1816 | 83 | |
| GenBank mRNA | AF301152.1 | - | 16 - 17 | 1816 | 83 | |
| AK138940.1 | - | 2 - 3 | 240 | 83 | ||
Set Membership: |
|