Search
Search
| Gene Symbol: Irf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018533.1 | - | 6 - 7 | 659 | 64 | |
| AK036488.1 | - | 6 - 7 | 659 | 64 | ||
| AK137450.1 | - | 6 - 7 | 1194 | 64 | ||
| AK146004.1 | - | 6 - 7 | 659 | 64 | ||
| AK150320.1 | - | 6 - 7 | 660 | 64 | ||
| AK150370.1 | - | 6 - 7 | 659 | 64 | ||
| AK151324.1 | - | 6 - 7 | 659 | 64 | ||
| AK151480.1 | - | 6 - 7 | 659 | 64 | ||
| AK152477.1 | - | 6 - 7 | 659 | 64 | ||
| AK152906.1 | - | 6 - 7 | 659 | 64 | ||
| AK153068.1 | - | 6 - 7 | 659 | 64 | ||
| AK153166.1 | - | 6 - 7 | 659 | 64 | ||
| AK153395.1 | - | 6 - 7 | 659 | 64 | ||
| AK154662.1 | - | 6 - 7 | 659 | 64 | ||
| AK156481.1 | - | 6 - 7 | 659 | 64 | ||
| AK157147.1 | - | 6 - 7 | 659 | 64 | ||
| AK157170.1 | - | 6 - 7 | 659 | 64 | ||
| AK157249.1 | - | 6 - 7 | 659 | 64 | ||
| AK157700.1 | - | 6 - 7 | 658 | 64 | ||
| AK159835.1 | - | 6 - 7 | 659 | 64 | ||
| BC005450.1 | - | 6 - 7 | 640 | 3487214 | 64 | |
| CT010225.1 | - | 5 - 6 | 605 | 64 | ||
| M32489.1 | - | 6 - 7 | 625 | 64 | ||
Set Membership: |
|