Search
Search

Galns| Gene Symbol: Galns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193645.1 | NP_001180574.1 | 8 - 9 | 1046 | 70 | |
| NM_016722.4 | NP_057931.3 | 8 - 9 | 1046 | 70 | ||
| GenBank mRNA | AF111346.1 | - | 8 - 9 | 967 | 70 | |
| AK033946.1 | - | 8 - 9 | 964 | 70 | ||
| AK159592.1 | - | 8 - 9 | 1023 | 70 | ||
| AK220245.1 | - | 8 - 9 | 948 | 70 | ||
| BC004002.1 | - | 8 - 9 | 900 | 3492664 | 70 | |
| CB600418.1 | - | 5 - 6 | 507 | 70 | ||
Set Membership: |
|