Search
Search

Pla2g15| Gene Symbol: Pla2g15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133792.2 | NP_598553.1 | 4 - 5 | 595 | 67 | |
| GenBank mRNA | AF468958.1 | - | 4 - 5 | 507 | 67 | |
| AK085194.1 | - | 4 - 5 | 584 | 67 | ||
| AK155004.1 | - | 4 - 5 | 591 | 67 | ||
| AK163111.1 | - | 4 - 5 | 584 | 67 | ||
| AK170814.1 | - | 4 - 5 | 583 | 67 | ||
| AK190514.1 | - | 1 - 2 | 67 | |||
| BC019373.1 | - | 4 - 5 | 595 | 3964729 | 67 | |
Set Membership: |
|