Search
Search

Itga2b| Gene Symbol: Itga2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010575.2 | NP_034705.2 | 19 - 20 | 2112 | 83 | |
| GenBank mRNA | AF170316.1 | - | 19 - 20 | 1973 | 83 | |
| AK048700.1 | - | 14 - 15 | 1620 | 83 | ||
| AK140749.1 | - | 14 - 15 | 1764 | 83 | ||
| AK142289.1 | - | 19 - 20 | 1974 | 83 | ||
| AK154619.1 | - | 19 - 20 | 2113 | 83 | ||
| AK156872.1 | - | 18 - 19 | 2294 | 83 | ||
| AK157012.1 | - | 18 - 19 | 2297 | 83 | ||
| AK158443.1 | - | 19 - 20 | 1964 | 83 | ||
| BC120493.1 | - | 19 - 20 | 1962 | 40129416 | 83 | |
| BC137570.1 | - | 19 - 20 | 1962 | 8860589 | 83 | |
Set Membership: |
|