Search
Search

Ces2c| Gene Symbol: Ces2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145603.2 | NP_663578.1 | 1 - 2 | 142 | 56 | |
| GenBank mRNA | AB110073.1 | - | 1 - 2 | 142 | 56 | |
| AK136411.1 | - | 1 - 2 | 141 | 56 | ||
| BC015290.1 | - | 1 - 2 | 126 | 4241028 | 56 | |
| BC024552.1 | - | 1 - 2 | 124 | 4990820 | 56 | |
| BC031170.1 | - | 1 - 2 | 124 | 4984806 | 56 | |
| BC034178.1 | - | 1 - 2 | 111 | 4971285 | 56 | |
| BC034180.1 | - | 1 - 2 | 124 | 4972755 | 56 | |
| BC034191.1 | - | 1 - 2 | 124 | 4982995 | 56 | |
Set Membership: |
|