Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Psmd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025894.2 | NP_080170.1 | 8 - 9 | 956 | 74 | |
GenBank mRNA | AK005114.1 | - | 8 - 9 | 990 | 74 | |
AK007619.1 | - | 8 - 9 | 1001 | 74 | ||
AK007680.1 | - | 8 - 9 | 930 | 74 | ||
AK008646.1 | - | 8 - 9 | 955 | 74 | ||
AK009969.1 | - | 8 - 9 | 959 | 74 | ||
AK017843.1 | - | 8 - 9 | 973 | 74 | ||
AK150910.1 | - | 8 - 9 | 979 | 74 | ||
AK151242.1 | - | 8 - 9 | 957 | 74 | ||
AK152041.1 | - | 8 - 9 | 979 | 74 | ||
AK152240.1 | - | 8 - 9 | 988 | 74 | ||
AK162779.1 | - | 957 | 74 | |||
AK167236.1 | - | 8 - 9 | 964 | 74 | ||
AK209461.1 | - | 1 - 2 | 43 | 74 | ||
BC004694.1 | - | 8 - 9 | 974 | 3498952 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |