Search
Search

Ergic3| Gene Symbol: Ergic3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025516.4 | NP_079792.1 | 6 - 7 | 640 | 83 | |
| GenBank mRNA | AK009350.1 | - | 6 - 7 | 641 | 83 | |
| AK050285.1 | - | 6 - 7 | 655 | 83 | ||
| AK190333.1 | - | 2 - 3 | 216 | 83 | ||
| BC043720.1 | - | 6 - 7 | 665 | 5375947 | 83 | |
| BC057130.1 | - | 6 - 7 | 659 | 6819420 | 83 | |
Set Membership: |
|