Search
Search

St6galnac1| Gene Symbol: St6galnac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011371.2 | NP_035501.2 | 3 - 4 | 884 | 59 | |
| GenBank mRNA | BC141095.1 | - | 867 | 9055707 | 59 | |
| BC145373.1 | - | 867 | 9053931 | 59 | ||
| Y11274.1 | - | 3 - 4 | 884 | 59 | ||
Set Membership: |
|