Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01253206_m1
          See other NRP1 GE Assays ›
          Gene Symbol:
          Nrp1
          Gene Name:
          neuropilin 1
          Gene Aliases:
          C530029I03, NP-1, NPN-1, Npn1, Nrp
          Chromosome Location:
          Chr.8: 128358285 - 128505476 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01253206_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nrp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008737.2 NP_032763.2 13 - 14 2545 87
          GenBank mRNA AK141964.1 - 13 - 14 2337 87
          AK148372.1 - 13 - 14 2521 87
          AK149752.1 - 8 - 9 1127 87
          AK159219.1 - 13 - 14 2337 87
          BC060129.1 - 13 - 14 2325 6409596 87
          D50086.1 - 13 - 14 2405 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001594.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nrp1

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          patterning of blood vessels
          neuron migration
          sprouting angiogenesis
          cell migration involved in sprouting angiogenesis
          multicellular organism development
          nervous system development
          axon guidance
          axonal fasciculation
          heart development
          motor neuron axon guidance
          organ morphogenesis
          dendrite development
          cell migration
          peptidyl-tyrosine phosphorylation
          facial nerve structural organization
          trigeminal nerve morphogenesis
          trigeminal nerve structural organization
          vestibulocochlear nerve structural organization
          nerve development
          branchiomotor neuron axon guidance
          gonadotrophin-releasing hormone neuronal migration to the hypothalamus
          cell differentiation
          negative regulation of axon extension
          retinal ganglion cell axon guidance
          cellular response to hepatocyte growth factor stimulus
          endothelial cell chemotaxis
          cellular response to vascular endothelial growth factor stimulus
          ventral trunk neural crest cell migration
          vascular endothelial growth factor signaling pathway
          VEGF-activated neuropilin signaling pathway
          otic placode formation
          negative regulation of neuron apoptotic process
          platelet-derived growth factor receptor signaling pathway
          vascular endothelial growth factor receptor signaling pathway
          hepatocyte growth factor receptor signaling pathway
          sympathetic nervous system development
          neuron development
          regulation of axon extension involved in axon guidance
          positive regulation of axon extension involved in axon guidance
          negative regulation of axon extension involved in axon guidance
          artery morphogenesis
          axon extension involved in axon guidance
          positive regulation of peptidyl-tyrosine phosphorylation
          positive chemotaxis
          axonogenesis involved in innervation
          dichotomous subdivision of terminal units involved in salivary gland branching
          angiogenesis involved in coronary vascular morphogenesis
          cell migration involved in coronary vasculogenesis
          coronary artery morphogenesis
          retina vasculature morphogenesis in camera-type eye
          renal artery morphogenesis
          sympathetic ganglion development
          trigeminal ganglion development
          ganglion morphogenesis
          positive regulation of ERK1 and ERK2 cascade
          semaphorin-plexin signaling pathway
          commissural neuron axon guidance
          regulation of retinal ganglion cell axon guidance
          sensory neuron axon guidance
          sympathetic neuron projection extension
          sympathetic neuron projection guidance
          neural crest cell migration involved in autonomic nervous system development
          toxin transport
          semaphorin-plexin signaling pathway involved in neuron projection guidance
          semaphorin-plexin signaling pathway involved in axon guidance
          positive regulation of retinal ganglion cell axon guidance
          VEGF-activated neuropilin signaling pathway involved in axon guidance
          protein localization to early endosome
          facioacoustic ganglion development
          negative regulation of extrinsic apoptotic signaling pathway
          vascular endothelial growth factor-activated receptor activity
          protein binding
          heparin binding
          semaphorin receptor activity
          growth factor binding
          vascular endothelial growth factor binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800013071.mm.1 NM_008737.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013071.mm.1 NM_008737.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013071.mm.1 TC0800001594.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline