Search
Search

Itgb1| Gene Symbol: Itgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010578.2 | NP_034708.1 | 6 - 7 | 1094 | 101 | |
| GenBank mRNA | AK076526.1 | - | 2 - 3 | 391 | 101 | |
| AK088016.1 | - | 6 - 7 | 995 | 101 | ||
| AK088729.1 | - | 6 - 7 | 841 | 101 | ||
| AK135521.1 | - | 5 - 6 | 770 | 101 | ||
| AK159689.1 | - | 6 - 7 | 936 | 101 | ||
| AK167682.1 | - | 6 - 7 | 992 | 101 | ||
| BC050906.1 | - | 6 - 7 | 967 | 5721348 | 101 | |
| X15202.1 | - | 5 - 6 | 787 | 101 | ||
| Y00769.1 | - | 6 - 7 | 882 | 101 | ||
Set Membership: |
|