Search
Search

Notum| Gene Symbol: Notum | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175263.4 | NP_780472.3 | 10 - 11 | 1233 | 88 | |
| GenBank mRNA | AK019987.1 | - | 10 - 11 | 1233 | 88 | |
| BC025832.1 | - | 5 - 6 | 703 | 5125723 | 88 | |
Set Membership: |
|