Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01253364_m1
          See other SRR GE Assays ›
          Gene Symbol:
          Srr
          Gene Name:
          serine racemase
          Gene Aliases:
          Srs
          Chromosome Location:
          Chr.11: 74905047 - 74925798 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01253364_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Srr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013761.4 NP_038789.1 1 - 2 180 69
          GenBank mRNA AB232340.1 - 1 - 2 132 69
          AK031687.1 - 1 - 2 52 69
          AK043738.1 - 1 - 2 86 69
          AK080830.1 - 1 - 2 59 69
          AK170096.1 - 1 - 2 66 69
          AK170471.1 - 1 - 2 66 69
          BC011164.1 - 1 - 2 57 4195695 69

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AB232340.1, AK031687.1, AK043738.1, AK170096.1, BC011164.1, NM_013761.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Srr

          Panther Classification:

          Molecular Function -

          epimerase/racemase isomerase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular amino acid metabolic process
          L-serine metabolic process
          brain development
          metabolic process
          serine family amino acid metabolic process
          pyruvate biosynthetic process
          protein homotetramerization
          D-serine metabolic process
          D-serine biosynthetic process
          nucleotide binding
          magnesium ion binding
          catalytic activity
          L-serine ammonia-lyase activity
          calcium ion binding
          protein binding
          ATP binding
          D-serine ammonia-lyase activity
          glycine binding
          lyase activity
          isomerase activity
          threonine racemase activity
          PDZ domain binding
          pyridoxal phosphate binding
          serine racemase activity
          protein homodimerization activity
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100029552.mm.1 NM_001163311.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029552.mm.1 NM_001163311.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029552.mm.1 NM_013761.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029552.mm.1 NM_013761.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029552.mm.1 TC1100003207.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100029559.mm.1 NM_001163311.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029559.mm.1 NM_001163311.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029559.mm.1 NM_013761.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029559.mm.1 NM_013761.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029559.mm.1 TC1100003207.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100029558.mm.1 NM_001163311.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029558.mm.1 NM_001163311.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029558.mm.1 NM_013761.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029558.mm.1 NM_013761.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029558.mm.1 TC1100003207.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100029546.mm.1 NM_013761.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029546.mm.1 NM_013761.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100029546.mm.1 TC1100003207.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 842789 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline