Search
Search

Mrps9| Gene Symbol: Mrps9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023514.4 | NP_076003.3 | 1 - 2 | 192 | 100 | |
| GenBank mRNA | AK009082.1 | - | 1 - 2 | 142 | 100 | |
| AK031497.1 | - | 1 - 2 | 141 | 100 | ||
| AK049465.1 | - | 1 - 2 | 150 | 100 | ||
| AK144656.1 | - | 1 - 2 | 150 | 100 | ||
| AK150985.1 | - | 1 - 2 | 150 | 100 | ||
| AK214977.1 | - | 1 - 2 | 30 | 100 | ||
| BC116437.2 | - | 1 - 2 | 142 | 40104493 | 100 | |
Set Membership: |
|