Search
Search

Creb1| Gene Symbol: Creb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009952.2 | NP_034082.1 | 4 - 5 | 523 | 125 | |
| GenBank mRNA | AF448507.1 | - | 4 - 5 | 399 | 125 | |
| AF448508.1 | - | 1051 | 125 | |||
| M95106.1 | - | 4 - 5 | 433 | 125 | ||
Set Membership: |
|